Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130145.2 | NP_001123617.1 | 9 | 3558 | ||
NM_001195044.1 | NP_001181973.1 | 8 | 3510 | |||
NM_001195045.1 | NP_001181974.1 | 9 | 3155 | |||
NM_001282097.1 | NP_001269026.1 | 8 | 3444 | |||
NM_001282098.1 | NP_001269027.1 | 7 | 3408 | |||
NM_001282099.1 | NP_001269028.1 | 8 | 3456 | |||
NM_001282100.1 | NP_001269029.1 | 8 | 3522 | |||
NM_001282101.1 | NP_001269030.1 | 9 | 3570 | |||
NM_006106.4 | NP_006097.2 | 7 | 3396 | |||
XM_005271378.3 | XP_005271435.1 | 9 | 4080 | |||
XM_005271380.3 | XP_005271437.1 | 9 | 4068 | |||
XM_005271381.3 | XP_005271438.1 | 8 | 4032 | |||
XM_005271383.3 | XP_005271440.1 | 8 | 4020 | |||
XM_011542555.2 | XP_011540857.1 | 9 | 3168 | |||
XM_011542556.2 | XP_011540858.1 | 9 | 3162 | |||
XM_017017093.1 | XP_016872582.1 | 8 | 3102 | |||
GenBank mRNA | AL832620.2 | - | 8 | 2851 | ||
BC026212.2 | - | 3 | 2139 |