Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031666.1 | NP_001026836.1 | 5 | 819 | ||
NM_001031809.1 | NP_001026979.1 | 6 | 990 | |||
NM_006138.4 | NP_006129.4 | 7 | 1128 | |||
XM_011545363.2 | XP_011543665.1 | 6 | 875 | |||
GenBank mRNA | BC008487.1 | - | 7 | 1073 | 4102067 | |
L35848.1 | - | 7 | 1106 |