Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143835.1 | NP_001137307.1 | 24 | 3161 | ||
NM_006165.3 | NP_006156.2 | 22 | 3156 | |||
XM_005271567.3 | XP_005271624.1 | 24 | 3197 | |||
XM_005271568.3 | XP_005271625.1 | 23 | 3112 | |||
XM_011542850.2 | XP_011541152.1 | 25 | 3234 | |||
XM_011542851.2 | XP_011541153.1 | 24 | 3216 | |||
XM_011542852.2 | XP_011541154.1 | 23 | 3054 | |||
XM_017017793.1 | XP_016873282.1 | 23 | 3108 | |||
XM_017017794.1 | XP_016873283.1 | 23 | 3072 | |||
XM_017017795.1 | XP_016873284.1 | 25 | 3198 | |||
XM_017017796.1 | XP_016873285.1 | 23 | 3076 | |||
XM_017017799.1 | XP_016873288.1 | 25 | 3308 | |||
XM_017017800.1 | XP_016873289.1 | 25 | 3248 | |||
XM_017017801.1 | XP_016873290.1 | 10 | 2208 | |||
GenBank mRNA | AL512730.1 | - | 6 | 999 | ||
BC063280.1 | - | 24 | 3107 | 5271534 | ||
X80878.1 | - | 22 | 3165 |