Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256339.1 | NP_001243268.1 | 3 | 2486 | ||
NM_006167.3 | NP_006158.2 | 2 | 2759 | |||
NR_046072.1 | 1 | 1946 | ||||
GenBank mRNA | AF247704.1 | - | 3 | 2771 |