Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006191.2 | NP_006182.2 | 8 | 1084 | ||
GenBank mRNA | AK291796.1 | - | 8 | 837 | ||
BC001951.1 | - | 8 | 784 | 2823726 | ||
BC007561.1 | - | 8 | 798 | 2988781 | ||
BC019222.1 | - | 9 | 757 | |||
BC032111.2 | - | 8 | 765 | |||
BC069786.1 | - | 8 | 737 | 7262559 | ||
BC072007.1 | - | 8 | 764 | |||
U59435.1 | - | 8 | 762 | |||
U87954.1 | - | 8 | 860 |