Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317836.1 | NP_001304765.1 | 11 | 1162 | ||
NM_006315.5 | NP_006306.2 | 10 | 1063 | |||
GenBank mRNA | AK057124.1 | - | 10 | 2000 | ||
AK125801.1 | - | 11 | 2007 | |||
AK225892.1 | - | 11 | 1062 | |||
AK300290.1 | - | 11 | 1071 | |||
BC107061.2 | - | 8 | 688 | 40006331 | ||
BX648409.1 | - | 6 | 1411 | |||
CA454695.1 | - | 458 |