Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317836.1 | NP_001304765.1 | 12 | 2995 | ||
NM_006315.5 | NP_006306.2 | 11 | 2896 | |||
GenBank mRNA | AK125801.1 | - | 13 | 3572 | ||
BC050044.1 | - | 1 | 1020 | |||
BX640894.1 | - | 1607 |