Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006446.4 | NP_006437.3 | 15 | 2524 | ||
GenBank mRNA | AF205071.1 | - | 15 | 2554 | ||
AJ132573.1 | - | 14 | 2483 |