Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGRRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006568.2 | NP_006559.1 | 6 | 867 | ||
XM_011536362.2 | XP_011534664.1 | 6 | 756 | |||
XM_017020931.1 | XP_016876420.1 | 7 | 958 | |||
XM_017020932.1 | XP_016876421.1 | 7 | 824 | |||
GenBank mRNA | AK090727.1 | - | 2 | 1856 | ||
AK289478.1 | - | 6 | 898 | |||
AK315318.1 | - | 6 | 908 | |||
BC015063.1 | - | 6 | 833 | 4245551 | ||
U66469.1 | - | 6 | 803 |