Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113178.2 | NP_001106649.1 | 4 | 1134 | ||
NM_006599.3 | NP_006590.1 | 3 | 1080 | |||
NM_138713.3 | NP_619727.2 | 4 | 1134 | |||
NM_138714.3 | NP_619728.2 | 5 | 1196 | |||
NM_173214.2 | NP_775321.1 | 4 | 1142 | |||
NM_173215.2 | NP_775322.1 | 4 | 1142 | |||
XM_006721125.3 | XP_006721188.1 | 3 | 1805 | |||
XM_011522817.2 | XP_011521119.1 | 3 | 1080 | |||
XM_011522818.2 | XP_011521120.1 | 6 | 1269 | |||
XM_017022870.1 | XP_016878359.1 | 3 | 1429 | |||
XM_017022871.1 | XP_016878360.1 | 4 | 1156 | |||
GenBank mRNA | AB020634.1 | - | 3 | 1062 | ||
AF089824.1 | - | 4 | 1084 | |||
AF134870.1 | - | 1 | 516 | |||
AF163836.1 | - | 2 | 603 | |||
AF346509.1 | - | 5 | 2082 | |||
AJ005683.1 | - | 4 | 758 | |||
AJ005684.1 | - | 4 | 758 | |||
AJ243298.1 | - | 4 | 901 | |||
AJ243299.1 | - | 4 | 893 | |||
BC131509.1 | - | 4 | 1038 | 40083218 | ||
BC146765.1 | - | 3 | 1062 | 9007208 |