Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282621.1 | NP_001269550.1 | 2 | 1371 | ||
NM_006667.4 | NP_006658.1 | 3 | 1527 | |||
GenBank mRNA | AK293618.1 | - | 2 | 1291 | ||
BC034238.1 | - | 3 | 1461 | 3863377 | ||
Y12711.2 | - | 3 | 1486 |