Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257180.1 | NP_001244109.1 | 11 | 3133 | ||
NM_001257181.1 | NP_001244110.1 | 11 | 2764 | |||
NM_006749.4 | NP_006740.1 | 11 | 3096 | |||
XM_005273613.3 | XP_005273670.1 | 11 | 2871 | |||
XM_005273615.3 | XP_005273672.1 | 12 | 2947 | |||
XM_006716390.3 | XP_006716453.1 | 10 | 2730 | |||
XM_006716391.3 | XP_006716454.1 | 12 | 6351 | |||
XM_017013748.1 | XP_016869237.1 | 11 | 2987 | |||
XM_017013749.1 | XP_016869238.1 | 10 | 6075 | |||
XM_017013750.1 | XP_016869239.1 | 12 | 3006 | |||
XM_017013751.1 | XP_016869240.1 | 12 | 2880 | |||
XM_017013752.1 | XP_016869241.1 | 12 | 3122 | |||
GenBank mRNA | BC028600.2 | - | 11 | 2808 | 4820601 | |
L20852.1 | - | 10 | 2682 |