Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278426.3 | NP_001265355.2 | 12 | 3284 | ||
NM_001278427.3 | NP_001265356.2 | 12 | 3281 | |||
NM_001278428.3 | NP_001265357.2 | 12 | 3284 | |||
NM_001278429.3 | NP_001265358.2 | 11 | 3251 | |||
GenBank mRNA | BC026309.1 | - | 12 | 3103 | 4795550 |