Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001997.4 | NP_001988.1 | 2, 3 | 202 | ||
GenBank mRNA | AK026639.1 | - | 2, 3 | 187 | ||
BC033877.1 | - | 2, 3 | 120 | 5758816 | ||
BP296770.1 | - | 2, 3 | 190 | |||
BU602831.1 | - | 2, 3 | 205 | |||
CR541974.1 | - | 1, 2 | 83 | |||
X65923.1 | - | 2, 3 | 139 |