Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276457.1 | NP_001263386.1 | 6 | 2676 | ||
NM_001276458.1 | NP_001263387.1 | 7 | 2727 | |||
NM_001276459.1 | NP_001263388.1 | 6 | 2640 | |||
NM_006874.3 | NP_006865.1 | 7 | 2763 | |||
NM_201999.2 | NP_973728.1 | 9 | 2913 | |||
XM_005262803.2 | XP_005262860.1 | 10 | 3222 | |||
XM_005262804.2 | XP_005262861.1 | 10 | 3186 | |||
XM_005262805.2 | XP_005262862.1 | 9 | 3099 | |||
XM_006714128.2 | XP_006714191.1 | 9 | 3135 | |||
XM_006714129.3 | XP_006714192.1 | 9 | 3001 | |||
XM_011531710.2 | XP_011530012.1 | 10 | 3179 | |||
XM_011531711.2 | XP_011530013.1 | 8 | 2728 | |||
XM_011531712.2 | XP_011530014.1 | 7 | 2719 | |||
XM_011531714.2 | XP_011530016.1 | 4 | 2196 | |||
XM_017007856.1 | XP_016863345.1 | 9 | 3021 | |||
XM_017007857.1 | XP_016863346.1 | 9 | 2965 | |||
XM_017007858.1 | XP_016863347.1 | 8 | 2970 | |||
XM_017007859.1 | XP_016863348.1 | 9 | 3056 | |||
XM_017007860.1 | XP_016863349.1 | 8 | 2878 | |||
XM_017007861.1 | XP_016863350.1 | 9 | 3006 | |||
GenBank mRNA | U43188.1 | - | 9 | 2891 | ||
U43189.1 | - | 7 | 2662 |