Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006893.2 | NP_008824.2 | 6 | 910 | ||
XM_005277509.3 | XP_005277566.1 | 6 | 825 | |||
XM_006711196.3 | XP_006711259.1 | 6 | 781 | |||
XM_011509257.2 | XP_011507559.1 | 6 | 825 | |||
XM_011509258.2 | XP_011507560.1 | 6 | 825 | |||
XM_017000532.1 | XP_016856021.1 | 6 | 781 | |||
GenBank mRNA | AB209364.1 | - | 1 | 468 | ||
AF220417.2 | - | 6 | 818 | |||
AK001852.1 | - | 6 | 823 | |||
AK225259.1 | - | 6 | 804 | |||
AL162001.1 | - | 1 | 116 | |||
BC001585.1 | - | 6 | 808 | 3463026 | ||
BC039134.1 | - | 6 | 761 | 4339127 | ||
BC058905.1 | - | 6 | 775 | 6067337 |