Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 7 | 678 | ||
NM_001278283.1 | NP_001265212.1 | 8 | 758 | |||
NM_001278285.1 | NP_001265214.1 | 5 | 586 | |||
NM_001278286.1 | NP_001265215.1 | 6 | 602 | |||
NM_001278288.1 | NP_001265217.1 | 5 | 593 | |||
NM_001278289.1 | NP_001265218.1 | 7 | 742 | |||
NM_001318926.1 | NP_001305855.1 | 7 | 727 | |||
NM_001318927.1 | NP_001305856.1 | 7 | 771 | |||
NM_001318928.1 | NP_001305857.1 | 7 | 622 | |||
NM_001318930.1 | NP_001305859.1 | 6 | 662 | |||
NM_001318931.1 | NP_001305860.1 | 6 | 624 | |||
NM_006916.2 | NP_008847.1 | 6 | 647 | |||
NM_199229.2 | NP_954699.1 | 6 | 673 | |||
XM_006712677.3 | XP_006712740.1 | 7 | 620 | |||
GenBank mRNA | AJ224326.1 | - | 5 | 415 | ||
AK056028.1 | - | 6 | 584 | |||
AK093658.1 | - | 8 | 689 | |||
AK096199.1 | - | 7 | 509 | |||
AK291035.1 | - | 6 | 600 | |||
AK303184.1 | - | 5 | 513 | |||
AK308741.1 | - | 6 | 511 | |||
AL833120.1 | - | 6 | 596 | |||
BC005148.2 | - | 6 | 581 | 3505010 | ||
BC016764.1 | - | 6 | 610 | 4107315 | ||
BC072401.1 | - | 6 | 582 | 6047438 | ||
BX403091.2 | - | 7 | 633 | |||
BX643719.1 | - | 7 | 697 | |||
CN372165.1 | - | 7 | 665 | |||
DA237285.1 | - | 5 | 520 | |||
DA336224.1 | - | 6 | 533 | |||
DA592494.1 | - | 7 | 550 |