Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276373.1 | NP_001263302.1 | 11 | 1463 | ||
NM_007122.4 | NP_009053.1 | 11 | 1559 | |||
NM_207005.2 | NP_996888.1 | 11 | 1528 | |||
GenBank mRNA | AL832119.1 | - | 11 | 1596 | ||
BC035505.2 | - | 11 | 1507 | 4559819 | ||
X55666.1 | - | 11 | 1472 |