Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108201
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
24
2591
NM_001178107.1
NP_001171578.1
23
2504
NM_001178108.1
NP_001171579.1
23
2498
NM_001178109.1
NP_001171580.1
22
2408
NM_001178110.1
NP_001171581.1
21
2318
NM_001178113.1
NP_001171584.1
15
2048
NM_001178115.1
NP_001171586.1
9
1399
NM_007150.3
NP_009081.2
23
2495
XM_005274730.2
XP_005274787.1
25
2698
XM_005274731.2
XP_005274788.1
25
2695
XM_005274735.2
XP_005274792.1
24
2608
XM_005274737.2
XP_005274794.1
24
2602
XM_005274738.2
XP_005274795.1
24
2599
XM_005274740.2
XP_005274797.1
23
2512
XM_005274741.2
XP_005274798.1
23
2512
XM_005274742.2
XP_005274799.1
23
2509
XM_005274744.2
XP_005274801.1
22
2425
XM_005274745.2
XP_005274802.1
22
2422
XM_005274746.3
XP_005274803.1
21
2341
XM_011531194.1
XP_011529496.1
23
2503
XM_011531195.1
XP_011529497.1
24
2500
XM_017029821.1
XP_016885310.1
26
2830
XM_017029822.1
XP_016885311.1
27
2850
XM_017029823.1
XP_016885312.1
26
2827
XM_017029824.1
XP_016885313.1
25
2746
XM_017029825.1
XP_016885314.1
25
2743
XM_017029826.1
XP_016885315.1
25
2740
XM_017029827.1
XP_016885316.1
25
2737
XM_017029828.1
XP_016885317.1
25
2734
XM_017029829.1
XP_016885318.1
24
2653
XM_017029830.1
XP_016885319.1
24
2647
XM_017029831.1
XP_016885320.1
24
2644
XM_017029832.1
XP_016885321.1
24
2635
XM_017029833.1
XP_016885322.1
23
2569
XM_017029834.1
XP_016885323.1
23
2557
XM_017029835.1
XP_016885324.1
23
2518
GenBank mRNA
AK056517.1
-
12
1554
AK095258.1
-
9
1371
AK296967.1
-
23
2498
AY997296.1
-
23
2484
BX537525.1
-
9
3073
Y09538.1
-
15
1776
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0