Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288985.1 | NP_001275914.1 | 19 | 2637 | ||
NM_001288986.1 | NP_001275915.1 | 18 | 2476 | |||
NM_007168.3 | NP_009099.1 | 17 | 2356 | |||
XM_005256938.2 | XP_005256995.1 | 18 | 2504 | |||
XM_005256940.4 | XP_005256997.1 | 17 | 2409 | |||
XM_011524191.2 | XP_011522493.1 | 18 | 2467 | |||
XM_011524192.2 | XP_011522494.1 | 18 | 2467 | |||
GenBank mRNA | AB020629.1 | - | 17 | 2315 | ||
AK295876.1 | - | 18 | 2449 | |||
AK304286.1 | - | 17 | 2308 | |||
AK304360.1 | - | 18 | 2447 | |||
BC037309.1 | - | 19 | 2404 | |||
BC130280.1 | - | 19 | 2486 | 40146311 |