Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007269.2 | NP_009200.2 | 6, 7 | 521 | ||
GenBank mRNA | AF032922.1 | - | 6, 7 | 503 | ||
AK223170.1 | - | 6, 7 | 500 | |||
AK290134.1 | - | 6, 7 | 505 | |||
AK291322.1 | - | 6, 7 | 501 | |||
AK312910.1 | - | 6, 7 | 498 | |||
BC047764.1 | - | 6, 7 | 496 | 5585614 | ||
D63506.1 | - | 6, 7 | 497 |