Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 8 | 1501 | ||
NM_001284343.1 | NP_001271272.1 | 4 | 579 | |||
NM_007270.4 | NP_009201.2 | 7 | 1342 | |||
GenBank mRNA | AB209352.1 | - | 1 | 726 | ||
AF089745.1 | - | 7 | 1013 | |||
AK075331.1 | - | 7 | 1306 | |||
AK128597.1 | - | 8 | 1283 | |||
AK293440.1 | - | 4 | 557 | |||
AK293954.1 | - | 6 | 915 | |||
AK294266.1 | - | 6 | 1126 | |||
AK300328.1 | - | 8 | 1464 | |||
AK304643.1 | - | 4 | 417 | |||
AK316290.1 | - | 2 | 232 | |||
BC007443.2 | - | 1 | 58 | |||
BC064418.1 | - | 2 | 265 | |||
BC101723.1 | - | 7 | 1270 | 8069229 | ||
BC112053.1 | - | 7 | 1270 | 8327521 | ||
BQ688269.1 | - | 4 | 579 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 3 | 321 | |||
GenBank mRNA | BM041005.1 | - | 3 | 322 |