Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 23 | 6280 | ||
NM_007297.3 | NP_009228.2 | 22 | 6188 | |||
NM_007298.3 | NP_009229.2 | 22 | 2755 | |||
NM_007299.3 | NP_009230.2 | 22 | 2856 | |||
NM_007300.3 | NP_009231.2 | 24 | 6343 | |||
NR_027676.1 | 23 | 6184 | ||||
GenBank mRNA | BC072418.1 | - | 22 | 2856 | ||
U68041.1 | - | 9 | 1159 |