Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 16 | 1843 | ||
NM_001164392.1 | NP_001157864.1 | 14 | 1752 | |||
NM_001164393.1 | NP_001157865.1 | 13 | 1642 | |||
NM_007358.3 | NP_031384.1 | 15 | 1923 | |||
XM_011541016.1 | XP_011539318.1 | 15 | 1879 | |||
XM_011541019.1 | XP_011539321.1 | 15 | 1671 | |||
XM_017000676.1 | XP_016856165.1 | 15 | 1778 | |||
XM_017000677.1 | XP_016856166.1 | 14 | 1708 | |||
XM_017000678.1 | XP_016856167.1 | 15 | 1661 | |||
XM_017000679.1 | XP_016856168.1 | 14 | 1755 | |||
GenBank mRNA | AF072814.1 | - | 14 | 1621 | ||
AF073293.1 | - | 5 | 1125 | |||
AF086908.1 | - | 14 | 1724 | |||
AJ010014.1 | - | 15 | 1874 | |||
AK290318.1 | - | 15 | 1875 | |||
AK302900.1 | - | 13 | 1594 | |||
BC010013.2 | - | 14 | 1676 | 4308565 |