Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007375.3 | NP_031401.1 | 6 | 2647 | ||
XM_017000863.1 | XP_016856352.1 | 6 | 2635 | |||
XM_017000864.1 | XP_016856353.1 | 6 | 2635 | |||
XM_017000865.1 | XP_016856354.1 | 6 | 2635 | |||
GenBank mRNA | AL045296.1 | - | 1 | 138 | ||
AL050265.1 | - | 6 | 2601 | |||
BC001487.2 | - | 6 | 2600 | |||
BC071657.1 | - | 6 | 2598 | 5498250 | ||
BC095435.1 | - | 6 | 2599 | 30389805 |