Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012086.4 | NP_036218.1 | 14 | 2058 | ||
XM_005246965.4 | XP_005247022.1 | 14 | 2062 | |||
GenBank mRNA | AF133123.1 | - | 14 | 1994 | ||
AK025560.1 | - | 14 | 1972 | |||
AK027050.1 | - | 8 | 1009 | |||
AK314718.1 | - | 14 | 1999 | |||
AL137453.1 | - | 11 | 1400 | |||
BC015995.1 | - | 6 | 757 | |||
BC043347.1 | - | 14 | 1913 | 5498609 |