Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012086.4 | NP_036218.1 | 18 | 2890 | ||
XM_005246965.4 | XP_005247022.1 | 18 | 2894 | |||
GenBank mRNA | AF085850.1 | - | 3 | 432 | ||
AF133123.1 | - | 18 | 2826 | |||
AK025560.1 | - | 18 | 2804 | |||
AK027050.1 | - | 12 | 1841 | |||
AL137453.1 | - | 15 | 2232 | |||
BC015995.1 | - | 10 | 1589 | |||
BC043347.1 | - | 18 | 2745 | 5498609 |