Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 7 | 1082 | ||
NM_001270365.1 | NP_001257294.1 | 5 | 881 | |||
NM_012190.3 | NP_036322.2 | 7 | 1184 | |||
NR_072979.1 | 7 | 1184 | ||||
XM_006713481.2 | XP_006713544.1 | 7 | 923 | |||
XM_011512355.1 | XP_011510657.1 | 7 | 916 | |||
XM_017005613.1 | XP_016861102.1 | 7 | 1307 | |||
XM_017005614.1 | XP_016861103.1 | 7 | 923 | |||
GenBank mRNA | AB209299.1 | - | 7 | 991 | ||
AF052732.1 | - | 7 | 944 | |||
AK222535.1 | - | 7 | 966 | |||
AK222694.1 | - | 7 | 966 | |||
AK294392.1 | - | 5 | 665 | |||
AK294530.1 | - | 6 | 733 | |||
AK307699.1 | - | 8 | 1095 | |||
AK308112.1 | - | 7 | 923 | |||
AL133015.1 | - | 5 | 551 | |||
BC027241.1 | - | 7 | 957 | 4484301 | ||
CR749807.1 | - | 7 | 1082 | |||
KU178540.1 | - | 6 | 834 | |||
KU178541.1 | - | 7 | 1018 | |||
KU178542.1 | - | 6 | 834 |