Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012205.2 | NP_036337.2 | 2 | 211 | ||
XM_005264230.3 | XP_005264287.1 | 2 | 1959 | |||
XM_011532729.2 | XP_011531031.1 | 2 | 1959 | |||
XM_011532730.2 | XP_011531032.1 | 3 | 305 | |||
XM_011532731.2 | XP_011531033.1 | 2 | 1959 | |||
XM_017003717.1 | XP_016859206.1 | 3 | 305 | |||
GenBank mRNA | AK295693.1 | - | 2 | 193 | ||
BC029510.1 | - | 2 | 194 | 5302724 | ||
BI602406.1 | - | 2 | 199 | |||
CR457063.1 | - | 2 | 136 | |||
DT215851.1 | - | 2 | 213 | |||
Z29481.1 | - | 2 | 176 |