Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012343.3 | NP_036475.3 | 7 | 1193 | ||
NM_182977.2 | NP_892022.2 | 7 | 1159 | |||
XM_005248274.4 | XP_005248331.1 | 7 | 1171 | |||
XM_005248275.4 | XP_005248332.1 | 1 | 162 | |||
XM_006714461.3 | XP_006714524.1 | 6 | 785 | |||
XM_011514001.2 | XP_011512303.1 | 7 | 1190 | |||
XM_011514002.2 | XP_011512304.1 | 6 | 880 | |||
XM_017009293.1 | XP_016864782.1 | 7 | 1061 | |||
GenBank mRNA | AL831822.2 | - | 7 | 1160 | ||
BC110543.1 | - | 7 | 1009 | 40036686 | ||
BC110544.1 | - | 6 | 1006 | 40036689 | ||
U40490.1 | - | 7 | 1081 | |||
Z50101.1 | - | 6 | 981 |