Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012343.3 | NP_036475.3 | 22 | 4089 | ||
NM_182977.2 | NP_892022.2 | 22 | 4055 | |||
XM_005248274.4 | XP_005248331.1 | 22 | 4067 | |||
XM_005248275.4 | XP_005248332.1 | 17 | 3317 | |||
XM_006714461.3 | XP_006714524.1 | 21 | 3681 | |||
XM_011514001.2 | XP_011512303.1 | 22 | 4086 | |||
XM_011514002.2 | XP_011512304.1 | 21 | 3776 | |||
XM_017009293.1 | XP_016864782.1 | 22 | 3957 | |||
GenBank mRNA | AL831822.2 | - | 22 | 4057 | ||
BC032370.1 | - | 6 | 1283 | |||
U40490.1 | - | 22 | 3977 |