Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284277.1 | NP_001271206.1 | 11 | 2521 | ||
NM_001284278.1 | NP_001271207.1 | 12 | 2585 | |||
NM_012399.4 | NP_036531.1 | 12 | 2607 | |||
XM_011530052.2 | XP_011528354.1 | 11 | 2534 | |||
GenBank mRNA | AK123384.1 | - | 4 | 1955 | ||
BC018704.1 | - | 12 | 2531 | 4643509 | ||
BC031427.1 | - | 11 | 2436 | 4341137 |