Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012417.3 | NP_036549.2 | 9 | 2224 | ||
NM_181671.2 | NP_858057.1 | 10 | 2343 | |||
XM_005257216.2 | XP_005257273.1 | 9 | 1724 | |||
XM_017024442.1 | XP_016879931.1 | 9 | 1335 | |||
XM_017024443.1 | XP_016879932.1 | 10 | 1454 | |||
XM_017024444.1 | XP_016879933.1 | 10 | 1845 | |||
XM_017024445.1 | XP_016879934.1 | 10 | 1467 | |||
GenBank mRNA | AK307138.1 | - | 7 | 1607 |