Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001204204.2 | NP_001191133.1 | 8 | 732 | ||
NM_001291932.1 | NP_001278861.1 | 9 | 905 | |||
NM_012429.4 | NP_036561.1 | 10 | 981 | |||
NM_033382.2 | NP_203740.1 | 10 | 981 | |||
GenBank mRNA | AB033012.2 | - | 10 | 880 | ||
AK056163.1 | - | 4 | 350 | |||
AK091402.1 | - | 8 | 768 | |||
AK223587.1 | - | 10 | 941 | |||
AK296540.1 | - | 9 | 795 | |||
AK303751.1 | - | 8 | 628 | |||
AK314390.1 | - | 10 | 914 | |||
AL096881.1 | - | 10 | 978 | |||
AY927434.1 | - | 5 | 315 | |||
AY927435.1 | - | LIT3468 AY927435 |
5 | 315 | ||
AY927436.1 | - | LIT3469 AY927436 |
5 | 315 | ||
AY927437.1 | - | LIT3470 AY927437 |
5 | 315 | ||
AY927438.1 | - | 8 | 603 | |||
AY927439.1 | - | 9 | 710 | |||
AY927441.1 | - | 5 | 315 | |||
BC058915.1 | - | 10 | 885 | 5178251 | ||
CR456571.1 | - | 10 | 878 |