Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002054.4 | NP_002045.1 | 2, 3 | 345 | ||
GenBank mRNA | BC005278.1 | - | 2, 3 | 188 | 3950435 | |
BM272442.1 | - | 2, 3 | 348 | |||
BT006813.1 | - | 1, 2 | 89 | |||
J04040.1 | - | 2, 3 | 126 |