Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 5 | 627 | ||
NM_001142557.1 | NP_001136029.1 | 2 | 400 | |||
NM_012484.2 | NP_036616.2 | 5 | 624 | |||
NM_012485.2 | NP_036617.2 | 4 | 579 | |||
GenBank mRNA | AF032862.1 | - | 5 | 478 | ||
AK290577.1 | - | 4 | 420 | |||
AK302975.1 | - | 6 | 434 | |||
AK303616.1 | - | 2 | 278 | |||
AK313603.1 | - | 5 | 545 | |||
AY623426.1 | - | 5 | 450 | |||
BC017793.1 | - | 4 | 448 | |||
BC030505.2 | - | 5 | 534 | |||
BC057235.1 | - | 4 | 549 | |||
BC070281.1 | - | 4 | 455 | |||
BC108904.1 | - | 5 | 479 | 40007903 | ||
U29343.1 | - | 5 | 505 |