Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013250.2 | NP_037382.2 | 3 | 910 | ||
XM_006718311.2 | XP_006718374.1 | 3 | 950 | |||
GenBank mRNA | AF056618.1 | - | 3 | 910 | ||
AK298365.1 | - | 3 | 870 | |||
AK302639.1 | - | 3 | 953 | |||
BC014538.1 | - | 1 | 412 | 3827937 |