Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013250.2 | NP_037382.2 | 7 | 2564 | ||
XM_006718311.2 | XP_006718374.1 | 7 | 2604 | |||
GenBank mRNA | AF056618.1 | - | 7 | 2567 |