Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301167.1 | NP_001288096.1 | 4 | 467 | ||
NM_013252.2 | NP_037384.1 | 5 | 512 | |||
XM_011515995.2 | XP_011514297.1 | 3 | 399 | |||
XM_017011915.1 | XP_016867404.1 | 4 | 470 | |||
XM_017011916.1 | XP_016867405.1 | 5 | 538 | |||
XM_017011917.1 | XP_016867406.1 | 4 | 469 | |||
GenBank mRNA | AF139768.1 | - | 5 | 466 | ||
AJ271684.1 | - | 5 | 512 | |||
AK292766.1 | - | 5 | 501 | |||
AK295200.1 | - | 4 | 404 | |||
AK313703.1 | - | 5 | 501 | |||
BC093714.1 | - | 4 | 336 | 7939559 | ||
BC112099.1 | - | 4 | 336 | 8327567 | ||
BC113098.1 | - | 5 | 454 | 40076723 | ||
BC113099.1 | - | 4 | 385 | 40076725 | ||
BC143392.1 | - | 3 | 267 |