Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 4 | 414 | ||
NM_001197317.2 | NP_001184246.1 | 3 | 303 | |||
NM_013269.5 | NP_037401.1 | 4 | 414 | |||
GenBank mRNA | AF133299.1 | - | 4 | 400 | ||
AF285087.1 | - | 4 | 392 | |||
AF285089.1 | - | 3 | 324 | |||
AK310150.1 | - | 4 | 485 | |||
AK313554.1 | - | 4 | 405 | |||
BC019883.1 | - | 4 | 394 | 4524891 | ||
BU929933.1 | - | 4 | 399 | |||
CB958277.1 | - | 4 | 411 | |||
DA814760.1 | - | 4 | 409 | |||
DB111977.1 | - | 4 | 415 | |||
DB124897.1 | - | 4 | 410 | |||
FN813350.1 | - | 3 | 276 | |||
KF958454.1 | - | 4 | 462 | |||
KF958455.1 | - | 4 | 387 | |||
KF958456.1 | - | 4 | 387 | |||
KF958457.1 | - | 4 | 462 | |||
KF958458.1 | - | 4 | 387 | |||
KF971856.1 | - | 4 | 387 | |||
KF971857.1 | - | 4 | 387 | |||
KF971858.1 | - | 3 | 276 |