Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002077.3 | NP_002068.1 | 3 | 443 | ||
XM_005251929.3 | XP_005251986.1 | 3 | 375 | |||
XM_006717062.3 | XP_006717125.1 | 3 | 439 | |||
XM_006717063.3 | XP_006717126.1 | 3 | 410 | |||
GenBank mRNA | AK307789.1 | - | 1 | 185 | ||
BC032853.1 | - | 3 | 413 | 4828793 | ||
BG699932.1 | - | 3 | 416 | |||
DA324844.1 | - | 3 | 445 | |||
U51587.1 | - | 3 | 435 |