Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101648.1 | NP_001095118.1 | 10, 11 | 2701 | ||
NM_013389.2 | NP_037521.2 | 10, 11 | 2701 | |||
XM_011515326.2 | XP_011513628.1 | 9, 10 | 2552 | |||
XM_011515327.2 | XP_011513629.1 | 10, 11 | 2956 | |||
XM_011515328.2 | XP_011513630.1 | 7, 8 | 2300 | |||
GenBank mRNA | AF192522.1 | - | 10, 11 | 2701 | ||
AY437865.1 | - | 10, 11 | 2645 | |||
AY515256.1 | - | 10, 11 | 2645 | |||
BC117178.1 | - | 10, 11 | 2699 | 40125729 | ||
FJ481111.1 | - | 10, 11 | 2645 |