Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 10, 11 | 1800 | ||
NM_182648.1 | NP_872589.1 | 10, 11 | 1800 | |||
XM_011536374.1 | XP_011534676.1 | 9, 10 | 1099 | |||
GenBank mRNA | AB032252.1 | - | 10, 11 | 1797 | ||
AB209255.1 | - | 2, 3 | 147 | |||
AF161435.1 | - | 10, 11 | 1487 | |||
AF213467.1 | - | 10, 11 | 1398 | |||
AF221130.1 | - | 9, 10 | 1232 |