Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002083.3 | NP_002074.2 | 1, 2 | 409 | ||
GenBank mRNA | BC005277.1 | - | 1, 2 | 249 | 3996732 | |
BC016756.1 | - | 1, 2 | 263 | 3681457 | ||
BC022820.1 | - | 1, 2 | 232 | 5429926 | ||
BC067221.1 | - | 1, 2 | 253 | 6668339 | ||
X68314.1 | - | 1, 2 | 264 |