Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014053.3 | NP_054772.1 | 8 | 1650 | ||
XM_011509446.2 | XP_011507748.1 | 7 | 1515 | |||
GenBank mRNA | AF118637.1 | - | 8 | 1650 | ||
AK001419.1 | - | 8 | 827 | |||
AK314481.1 | - | 8 | 1626 | |||
BC048312.1 | - | 8 | 1588 | 4417876 |