Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039847.2 | NP_001034936.1 | 3 | 346 | ||
NM_001039848.2 | NP_001034937.1 | 3 | 424 | |||
NM_002085.4 | NP_002076.2 | 3 | 346 | |||
GenBank mRNA | AK225859.1 | - | 3 | 265 | ||
BC010157.2 | - | 3 | 174 | |||
BC011836.2 | - | 3 | 267 | 3945177 | ||
BC021567.1 | - | 3 | 260 | 4581069 | ||
BC022071.1 | - | 3 | 283 | 4821595 | ||
BC032695.1 | - | 3 | 267 | 5227887 | ||
BC039849.1 | - | 3 | 335 | 5744771 | ||
BC046163.1 | - | 3 | 242 | 5742889 | ||
BC058329.1 | - | 3 | 263 | |||
BI561548.1 | - | 3 | 342 | |||
BM764133.1 | - | 3 | 425 | |||
BQ648717.1 | - | 3 | 211 | |||
CF127510.1 | - | 3 | 358 | |||
HY049693.1 | - | 3 | 347 | |||
X71973.1 | - | 3 | 308 |