Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014159.6 | NP_054878.5 | 3 | 2734 | ||
XM_011533631.2 | XP_011531933.1 | 4 | 4230 | |||
XM_011533632.2 | XP_011531934.1 | 3 | 2806 | |||
XM_011533633.2 | XP_011531935.1 | 4 | 4232 | |||
XM_011533634.1 | XP_011531936.1 | 3 | 2759 | |||
XM_017006270.1 | XP_016861759.1 | 4 | 4233 | |||
GenBank mRNA | AB051519.2 | - | 1 | 734 | ||
AJ238403.1 | - | 1 | 1581 | |||
AK026125.1 | - | 1 | 528 | |||
AK091413.1 | - | 1 | 1398 | |||
AK127782.1 | - | 1 | 2151 | |||
AK128096.1 | - | 3 | 2759 | |||
AK131371.1 | - | 3 | 2724 | |||
AL831959.1 | - | 1 | 1833 | |||
AY576987.1 | - | 1 | 1243 | |||
AY576988.1 | - | 1 | 1243 | |||
BC072440.1 | - | 1 | 1522 | |||
BC090954.1 | - | 1 | 524 | |||
BC117162.1 | - | 1 | 1357 | 40125713 | ||
BC117164.1 | - | 1 | 1357 | 40125715 | ||
BX649110.1 | - | 1 | 2296 |