Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014207.3 | NP_055022.2 | 11 | 2661 | ||
GenBank mRNA | BC027901.1 | - | 11 | 2620 | 5228913 |