Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014234.4 | NP_055049.1 | 9 | 890 | ||
GenBank mRNA | BC008185.1 | - | 9 | 885 | 4213813 | |
D82061.1 | - | 9 | 854 |