Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098517.1 | NP_001091987.1 | 8, 9 | 1263 | ||
NM_001301043.1 | NP_001287972.1 | 11, 12 | 1434 | |||
NM_001301044.1 | NP_001287973.1 | 10, 11 | 1380 | |||
NM_001301045.1 | NP_001287974.1 | 10, 11 | 1350 | |||
NM_014333.3 | NP_055148.3 | 9, 10 | 1347 | |||
XM_005271494.2 | XP_005271551.1 | 9, 10 | 1189 | |||
XM_017017457.1 | XP_016872946.1 | 11, 12 | 1393 | |||
XM_017017458.1 | XP_016872947.1 | 9, 10 | 1113 | |||
XM_017017459.1 | XP_016872948.1 | 12, 13 | 3387 | |||
XM_017017460.1 | XP_016872949.1 | 10, 11 | 3300 | |||
XM_017017461.1 | XP_016872950.1 | 9, 10 | 1230 | |||
GenBank mRNA | AF132811.1 | - | 9, 10 | 1221 | ||
AK075502.1 | - | 10, 11 | 1240 | |||
AK315245.1 | - | 8, 9 | 1196 | |||
AY309502.1 | - | 5, 6 | 448 | |||
AY358334.1 | - | 9, 10 | 1233 | |||
BC035930.1 | - | 9, 10 | 1239 | |||
BC125102.1 | - | 8, 9 | 1138 | |||
BC125103.1 | - | 8, 9 | 1137 | 40118330 | ||
BX641042.1 | - | 1, 2 | 8344 | |||
BX647110.1 | - | 2, 3 | 5029 | |||
CR457157.1 | - | 9, 10 | 1218 | |||
HE586496.1 | - | 9, 10 | 1221 | |||
HE586497.1 | - | 10, 11 | 1254 | |||
HE586498.1 | - | 11, 12 | 1308 | |||
HE586499.1 | - | 9, 10 | 1221 | |||
HE586500.1 | - | 10, 11 | 1304 | |||
HE586503.1 | - | 10, 11 | 1249 | |||
KJ534791.1 | - | 8, 9 | 1134 | |||
KJ534792.1 | - | 10, 11 | 1251 | |||
KJ534793.1 | - | 9, 10 | 1218 | |||
KJ534794.1 | - | 8, 9 | 1134 | |||
KJ534795.1 | - | 11, 12 | 1305 | |||
KJ534796.1 | - | 11, 12 | 1305 | |||
KJ534797.1 | - | 10, 11 | 1221 |